PUREZERO* LIMON* Nitrile Exam Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
LFS311LG LFS311MD LFS311SM LFS311XL LFS311XS
76554-006BX 59.11 CAD
76554-006 76554-034 76554-010 76554-020 76553-998
PUREZERO* LIMON* Nitrile Exam Gloves, Halyard
Gloves
Designed for use in laboratories, research environments, and clean areas with applications in pharmaceutical, medical device manufacturing, biotechnology, and food processing/handling.
  • Features a SMARTPULL* Dispenser to control dispensing and lessen waste
  • Powder-free and natural rubber latex-free
  • Compliant for food handling applications
  • AQL 1.0 (exceeds the ASTM standard of AQL 2.5 for critical defects or pinholes)
  • Ambidextrous
  • Beaded cuffs to prevent roll-down
Lab workers may spend hours every day dealing with a variety of chemicals, some with theMore Product Information