PUREZERO* ULTRA VIOLET* Nitrile Exam Gloves, Halyard
Supplier: O&M HALYARD, INC
LFS511LG LFS511MD LFS511SM LFS511XL LFS511XS
76554-030BX 55.11 CAD
76554-030 76554-038 76554-008 76554-026 76554-000
PUREZERO* ULTRA VIOLET* Nitrile Exam Gloves, Halyard
Gloves
Designed for use in laboratories, research environments, and clean areas with applications in pharmaceutical, medical device manufacturing, biotechnology, and food processing/handling.
  • Features a SMARTPULL* Dispenser to control dispensing and lessen waste
  • Powder-free and natural rubber latex-free
  • Compliant for food handling applications
  • AQL 1.0 (exceeds the ASTM standard of AQL 2.5 for critical defects or pinholes)
  • Ambidextrous
  • Beaded cuffs to prevent roll-down
Tested against 14 chemotherapy drugs, fentanyl, and 57 chemicals.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Cuff style Beaded
Finger thickness 3.5 mil
Palm thickness 2.8 mil
Sterility Non sterile