VWR® Cleanroom Tab Dividers

Supplier: VWR International
76547-916
76547-916PK 1001.26 CAD
76547-916
VWR® Cleanroom Tab Dividers
Binders and Indexes
11×17" cleanroom binder tab dividers.

  • Latex-free
  • Class 10 certified
  • Supplied 5 per pack

For use with cleanroom binders. These latex-free tab dividers are class 10 certified. They are tabbed and come in sets of 5 per pack.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR