API Thermo-Hydrometers
Supplier: Heathrow Scientific
6752HTS 6753HTS 6754HTS 6755HTS 6756HTS 6757HTS
76546-594EA 88.92 CAD
76546-594 76546-596 76546-598 76546-600 76546-602 76546-604
API Thermo-Hydrometers
Hydrometers
Our API scale hydrometers measure the scale which was selected in 1921 by the American Petroleum Institute (API), the U.S. Bureau of Mines, and the National Institute of Standards and Technology (NIST) as the standard hydrometer scale for petroleum products.
  • Safety blue thermometer liquid fill
The thermometer scale (where applicable) for these thermo-hydrometers is located in the body. All instruments are suitable for certification for use as a primary reference with NIST traceability.
The API scale is based on the following formula: °API at 60 °F =More Product Information