Hydrometer Cases
Supplier: Heathrow Scientific
66CS-13 66CS-13J 66CS-16 66CS-8 66CS-8F
76546-538EA 66.29 CAD
76546-538 76546-540 76546-542 76546-544 76546-546
Hydrometer Cases
Hydrometers
PVC hydrometer cases help protect hydrometers from damage. These cases feature a foam or felt padded insert, and padded bottom and cap.
  • Package includes white PVC case
Ideal for storing hydrometers when not in use.