Brix Hydrometers
Supplier: Heathrow Scientific
6601-1 6601-10 6601-11 6601-13 6601-14 6601-15 6601-2 6601-3 6601-4 6601-5
76546-270EA 50.14 CAD
76546-270 76546-272 76546-274 76546-276 76546-278 76546-280 76546-282 76546-284 76546-286 76546-288
Brix Hydrometers
Hydrometers
These brix scale hydrometers are calibrated to show the percentage of sucrose by weight at 20 °C. The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
  • Calibrated to show percent by weight
  • Wide variety of Brix ranges available
The data of the National Bureau of Standards are used.
Order Now
SPECIFICATIONS
Environmentally Preferable