VEE GEE Manual Polarimeter

Supplier: Heathrow Scientific
4601P
76546-244EA 2078.23 CAD
76546-244
VEE GEE Manual Polarimeter
Polarimeters
Our manual polarimeter features a full circular scale of ±180°.

  • Includes 100 and 200 mm sample cells with bubble trap
  • Rugged design and simple operation
  • Quality control and instructional applications
  • Wide measurement range of ±180°
  • Potential accuracy up to ±0.1°
  • LED light source

With the use of the Vernier scale, rotations can be measured to an accuracy of 0.1° (an experienced user can potentially achieve accuracy of 0.05°). The rugged design allows for heavy use at the instructional level.

Measurements are made by rotating the Vernier scale discMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR