Vee Gee Graduated Cylinders, Student Grade
Supplier: Heathrow Scientific
2355-100 2355-10 2355-25 2355-50
490011-084CS 247.82 CAD
490011-084 76546-192 76546-194 76546-196
Vee Gee Graduated Cylinders, Student Grade
Cylinders Measuring Cylinders
These high-quality graduated cylinders are made of premium borosilicate glass and comply with ASTM E-1272 standards, guaranteeing your investment in high-quality glassware.
  • Made out of high-quality borosilicate glass
  • Double metric scale
  • Graduated 'to contain'
  • Contains durable white enamel graduation markings
  • Feature hexagonal bases for added stability
  • All sizes come with a protective bumper guard
  • Calibration temperature: 20 °C (68 °F)
With their exceptional quality, precise accuracy, and ease of use, they are a great tool to use for all of your liquidMore Product Information