Vee Gee Volumetric Flasks with Glass Stopper
Supplier: Heathrow Scientific
2306A-2000 202GK-004 2306A-10 2306A-100 2306A-1000 2306A-200 2306A-25 2306A-250 2306A-5 2306A-50 2306A-500
490011-060CS 766.6 CAD
490011-060 76546-042 76546-128 76546-130 76546-132 76546-136 76546-138 76546-140 76546-142 76546-144 76546-146
Vee Gee Volumetric Flasks with Glass Stopper
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Wide range of sizes available (5 to 2000 ml)
Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity andMore Product Information