Vee Gee Volumetric Flasks with Snap Cap
Supplier: Heathrow Scientific
2303A-10 2303A-100 2303A-1000 2303A-200 2303A-25 2303A-250 2303A-50 2303A-500
76546-110CS 7007.79 CAD
76546-110 76546-112 76546-114 76546-116 76546-118 76546-120 76546-122 76546-124
Vee Gee Volumetric Flasks with Snap Cap
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for snap-cap fit.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 68 °F (20 °C)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a snap cap
  • Wide range of sizes available (10 to 1000 ml)
Our volumetric flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance toMore Product Information