Select Heated Ultrasonic Cleaners with 5 Modes and High Power, Elma
Supplier: Elma
1106996 1106997 1106998 1106999 1107000 1107001 1107002 1107003 1107004 1108777 1108778
76545-532EA 2020.25 CAD
76545-532 76545-534 76545-536 76545-538 76545-540 76545-542 76545-544 76545-546 76545-548 76545-550 76545-552
Select Heated Ultrasonic Cleaners with 5 Modes and High Power, Elma
Ultrasonic Cleaners
The Elmasonic Select series are powerful, versatile and programmable ultrasonic cleaners that allow users unparalleled control over cleaning and mixing. The 37 kHz units have five different modes.
  • Sweep mode - For uniform cleaning via a slight variation in ultrasonic frequency
  • Pulse mode - Brief spikes in power to remove stubborn contaminants or for mixing and sample prep
  • Dynamic mode - Combines sweep and pulse for optimized cleaning performance
  • Eco mode - For nearly silent, gentle cleaning
  • Degas mode - Quickly remove trapped air from solvents and fresh
More Product Information