Wall Cabinets, Blickman
Supplier: BLICKMAN INDUSTRIES
2020324000 2020224000 2020235000 2020335000 2020435000 2023035000 2020218000 2020218360 2020218480 2020224360 2020224480 2020235360 2020235480 2020247000 2020247360 2020247480 2020318000 2020318360 2020318480 2020324360 2020324480 2020335360 2020335480 2020347000 2020347360 2020347480 2020435360 2020435480 2020447000 2020447360 2020447480 2023035360 2023035480 2023047000 2023047360 2023047480
76545-000EA 5585.77 CAD
76545-000 76545-002 76545-004 76545-006 76545-008 76545-010 76545-090 76545-092 76545-094 76545-096 76545-098 76545-100 76545-102 76545-104 76545-106 76545-108 76545-110 76545-112 76545-114 76545-116 76545-118 76545-120 76545-122 76545-124 76545-126 76545-128 76545-130 76545-132 76545-134 76545-136 76545-138 76545-140 76545-142 76545-144 76545-146 76545-148
Wall Cabinets, Blickman
Furniture Storage Cabinets
Stainless steel wall cabinets and sloped tops are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.
  • Available with 13" depth, 18³/₄ to 47" width and 30 to 48" height
  • Designed for flush face construction with a recessed back panel for uneven wall surfaces
  • Solid stainless steel doors are double walled for added strength and sound deadened for optimal performance
  • Stainless steel and ¹/₄" tempered
More Product Information