Wall Shelves, Blickman
Supplier: BLICKMAN INDUSTRIES
1813612000 1813616000 1814812000 1814816000 1816012000 1816016000 1817212000 1817216000 1818412000
76545-024EA 1149.7 CAD
76545-024 76545-026 76545-028 76545-030 76545-032 76545-034 76545-036 76545-038 76545-040
Wall Shelves, Blickman
Furniture Shelves
Stainless steel wall shelves are constructed with quality 304 stainless steel, #4 polish and have a seamless welded face preventing dirt collection while providing long lasting durability through unparalleled strength and corrosion resistance.
  • 12 to 16" depth and 36 to 48" width standalone stainless steel wall shelf
  • Constructed for flush face wall surfaces
  • Supported by heavy-duty brackets and built for years of use
  • No sharp edges as front and sides of shelf form a channel, the back is turned up slightly and polished smooth
Standalone stainless steelMore Product Information