Cole-Parmer® SH-200 Orbital Shakers
Supplier: Antyila Scientific
51901-01 51901-11
76542-130EA 3440.13 CAD
76542-130 76542-132
Cole-Parmer® SH-200 Orbital Shakers
Shakers Orbital Shakers
Maintain consistent shaking speeds, even over long periods of time.
  • Smooth orbital shaking action with 16 mm orbit
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be used in incubators
These orbital shakers provide a smooth, uniform circular motion with a 16 mm orbit — ideal for larger samples and multiwell plates. Speed and time may be varied from 30 to 300 rpm. The controller is easy to use and features a timer and LED display. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shakerMore Product Information