VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Supplier: Ward's
H3710-HS-GM H3710-HS-P1-GM
76538-830EA 1228.29 CAD
76538-830 76538-832
VWR® GeneMate™ Digital Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
GeneMate™ Hotplate Magnetic Stirrers, 10×10" with temperature control up to 400 °C and stirring speeds up to 1500 rpm.
  • Excellent chemical resistance with white ceramic work surface
  • Stir volumes up to 15 L
  • Safety LED warns user when top plate is unsafe to touch
  • Choice of model with temperature probe for control over actual sample temperature
Certifications: CE marked.
Ordering information: 76538-832 is supplied with external temperature probe for control over actual sample temperature. Full specification details of stirrer and hotplate only modelsMore Product Information