High-Temperature Tube Furnaces for Horizontal and Vertical Operation up to 1800°C, Models RHTH and RHTV, Nabertherm
Supplier: NABERTHERM INC.
RH152V62AN-480 RH282V62AN-480 RH322V62AN-480 RH152V72AN-480 RH282V72AN-480 RH322V72AN-480 RH152V82AN-480 RH282V82AN-480 RH322V82AN-480 RV152V62AN-480 RV282V62AN-480 RV322V62AN-480 RV152V72AN-480 RV282V72AN-480 RV322V72AN-480 RV152V82AN-480 RV282V82AN-480 RV322V82AN-480
76538-436EA 47615.88 CAD
76538-436 76538-438 76538-440 76538-442 76538-444 76538-446 76538-448 76538-450 76538-452 76538-454 76538-456 76538-458 76538-460 76538-462 76538-464 76538-466 76538-468 76538-470
High-Temperature Tube Furnaces for Horizontal and Vertical Operation up to 1800°C, Models RHTH and RHTV, Nabertherm
Ovens Tube Furnaces
These high-temperature tube furnaces are available in either horizontal (type RHTH) or vertical (type RHTV) designs.
  • Tmax 1600, 1700 or 1800 °C
  • MoSi2 heating elements, mounted vertically for easy replacement
  • Insulation with vacuum-formed ceramic fiber plates
  • Rectangular outer housing with slots for convection cooling
  • Tube furnaces RHTV with frame for vertical operation
  • Housing made of sheets of textured stainless steel
  • Ceramic working tube made of material C 799 incl. fiber plugs operation under air
  • Type B thermocouple
  • Power unit with low-voltage
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable