Compact Tube Furnaces, Model R, Nabertherm
Supplier: NABERTHERM INC.
R-322L2AN-480 R-322O2AN-480 R-322W2AN3-480 R-322L3AN-480 R-322O3AN-480 R-322W3AN3-480 R-572L2AN-480 R-572O2AN-480 R-572W2AN3-480 R-572L3AN-480 R-572O3AN-480 R-572W3AN3-480 R-472L2AN-480 R-472O2AN-480 R-472W2AN3-480 R-472L3AN-480 R-472O3AN-480 R-472W3AN3-480 R-152L2AN-208 R-152L2AN-240 R-152O2AN-208 R-152O2AN-240 R-152L3AN-208 R-152L3AN-240 R-152O3AN-208 R-152O3AN-240 R-252L2AN-208 R-252L2AN-240 R-252O2AN-208 R-252O2AN-240 R-252W2AN3-208 R-252W2AN3-240 R-252L3AN-208 R-252L3AN-240 R-252O3AN-208 R-252O3AN-240 R-252W3AN3-208 R-252W3AN3-240
76538-344EA 16080.83 CAD
76538-344 76538-346 76538-348 76538-350 76538-352 76538-354 76538-356 76538-358 76538-360 76538-362 76538-364 76538-366 76538-368 76538-370 76538-372 76538-374 76538-376 76538-378 76538-380 76538-382 76538-384 76538-386 76538-388 76538-390 76538-392 76538-394 76538-396 76538-398 76538-400 76538-402 76538-404 76538-406 76538-408 76538-410 76538-412 76538-414 76538-416 76538-418
Compact Tube Furnaces, Model R, Nabertherm
Ovens Tube Furnaces
These compact tube furnaces with integrated control systems can be used universally for many processes.
  • Single-zoned design as standard; Three-zoned design extends the constant temperature accuracy within the heated length
  • Dual shell housing made of sheets of textured stainless steel
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Tmax 1200 °C: Type N thermocouple, Tmax 1300 °C: Type S thermocouple
  • Solid state relays provide for low noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
More Product Information