Muffle Furnaces with Scale and Software, Tmax 1100 °C, L 9/11 SW or LT 9/11 SW, Nabertherm
Supplier: NABERTHERM INC.
LW094K1LN1-208 LW094K1LN1-240 LW092K1TN1-208 LW092K1TN1-240 LW094H1LN1-208 LW094H1LN1-240 LW094H1ON1-208 LW094H1ON1-240
76538-166EA 27564.89 CAD
76538-166 76538-168 76538-170 76538-172 76538-182 76538-184 76538-186 76538-188
Muffle Furnaces with Scale and Software, Tmax 1100 °C, L 9/11 SW or LT 9/11 SW, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These weighing furnaces with an integrated precision scale and software, were designed especially for determining combustion loss in the laboratory. The determination of combustion loss is necessary, for instance, when analyzing sludges. Combustion loss is defined as the difference between the initial total mass and the combustion residue. During the process the software supplied records both temperature and weight loss.
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to
More Product Information