Muffle Furnaces, Models L and LT, Nabertherm
Supplier: NABERTHERM INC.
L-034H1ON1-120 L-034H1ON1-208 L-034K1ON1-120 L-034K1ON1-208 L-034K1ON1-240 L-054H1ON1-208 L-054H1ON1-240 L-054K1ON1-208 L-054K1ON1-240 L-094H1ON1-208 L-094H1ON1-240 L-094K1ON1-208 L-094K1ON1-240 L-154H1ON1-208 L-154H1ON1-240 L-154K1ON1-208 L-154K1ON1-240 L-244H1ON1-480 L-244K1ON1-480 L-404H1ON1-480 L-404K1ON1-480 L-404H1ON1-208 L-404K1ON1-208
75994-560EA 11220.09 CAD
75994-560 76537-868 76537-870 76537-874 76537-876 76537-878 76537-880 76537-882 76537-884 76537-886 76537-888 76537-890 76537-892 76537-894 76537-896 76537-898 76537-900 76537-902 76537-904 76537-906 76537-908 76537-910 76537-912 76537-914 CA76639-012 CA76639-016
Muffle Furnaces, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The muffle furnaces L 1/12 - LT 40/12 are the right choice for daily laboratory use.
  • Tmax 1100 °C or 1200 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for the 24/11 - LT 40/12 muffle furnaces)
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fiber material
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Optional fold down door (L models) which can
More Product Information