Drying Ovens, TR Series, Nabertherm
Supplier: NABERTHERM INC.
TR-905RN-480 TR-125RN-240 TR-245RN-240 TR-455RN-480 TR-065RN-240 TR-455LN-480 TR-245LN-240 TR-125LN-240 TR-905LN-480 TR-065LN-240 TR-065ON-240 TR-125ON-240 TR-245ON-240 TR-455ON-480 TR-905ON-480
75993-760EA 6862.94 CAD
75993-760 75993-762 75993-768 75993-772 75993-776 75994-522 75994-804 CA75993-760 CA75993-762 CA75993-768 CA75993-772 CA75993-776 76537-832 76537-834 76537-836 76537-838 76537-840 76537-842 76537-844 76537-846 76537-848 76537-850 76537-852 76537-854 76537-856 76537-858 76537-860 76537-862 76537-864 76537-866
Drying Ovens, TR Series, Nabertherm
Ovens Standard Ovens
With their maximum working temperature of up to 300 °C and air circulation, the ovens achieve nearly perfect temperature uniformity.
  • Tmax 300 °C
  • Working temperature range: + 5 °C above room temperature up to 300 °C
  • Ovens TR 60 - TR 240 designed as tabletop models
  • Ovens TR 450 and TR 1050 designed as floor standing models
  • Horizontal, air circulation results in temperature uniformity better than +/- 5 °C
  • Stainless steel chamber, alloy 304 (AISI)/(DIN material no. 1.4301), rust-resistant and easy to clean
  • Large handle to open and close the door
  • Charging in
More Product Information