Cole-Parmer® Essentials Graduated Square Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antyila Scientific
06251-00 06251-10 06251-20 06251-30 06251-40 06251-50
76534-266PK 51.63 CAD
76534-266 76534-268 76534-270 76534-272 76534-274 76534-276
Cole-Parmer® Essentials Graduated Square Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Resistant to acids, alcohols, and bases.
  • Drip proof spout for easy pouring
  • Space-saving square design
  • Autoclavable
0 to 135 °C.
Ordering information: Caps sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable