VWR® Mega Star 600 and 600R Small Bench Centrifuges and Packages
Supplier: VWR International
76533-880 76533-882 76533-884 76533-886
76533-880EA 4376.6 CAD
76533-880 76533-882 76533-884 76533-886
VWR® Mega Star 600 and 600R Small Bench Centrifuges and Packages
Centrifuges Benchtop Centrifuges
VWR® Mega Star 600 Ventilated and 600R Refrigerated Small Bench Centrifuge optimize bench space with maximized capacity in a compact footprint. Available in both a ventilated and refrigerated option, units come with a user-friendly interface and are adaptable in the ever evolving clinical and research laboratory.
  • Auto-Lock™ Rotor Exchange System: Units have secure, push-button Auto-Lock™ rotor exchange
  • Swap out rotors in as little as three seconds for trouble-free installation/removal and clear chamber access for cleaning convenience. No tools required
More Product Information
Order Now
SPECIFICATIONS
Model Mega Star 600 Mega Star 600R
Environmentally Preferable
Max. speed 4500 rpm (TX-150 rotor) 4500 rpm (TX-150 rotor)
Max. RCF 3260×g (TX-150 rotor) 3260×g (TX-150 rotor)
Refrigerated - Refrigerated