VWR® Micro Star 21 and 21R Microcentrifuges
Supplier: VWR International
76533-876 76533-878
76533-876EA 4250.2 CAD
76533-876 76533-878
VWR® Micro Star 21 and 21R Microcentrifuges
Centrifuges Microcentrifuges
VWR® Micro Star 21 ventilated and 21R refrigerated microcentrifuge with 24×1.5/2.0 ml rotor with ClickSeal™ biocontainment lid offer exceptional acceleration of up to 21000 xg in as little as 12 seconds.
  • Durable and flexible rotors
  • Outstanding corrosion resistance with lightweight engineered polymer rotors
  • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 ml tubes to mini-preps and spin columns
  • ClickSeal™ Biocontainment solution gives ergonomic one-click rotor opening and closing, as well as transparent design for added safety
More Product Information