Sentinel XT® Clear PAPR System with S-3112 Hood and Non-Incendive Blower, ILC Dover
Supplier: ILC DOVER LP
S-3320-01
76533-604EA 2544.99 CAD
76533-604
Sentinel XT® Clear PAPR System with S-3112 Hood and Non-Incendive Blower, ILC Dover
Respirators Respiratory Systems
The Sentinel XT® PAPR system offers up to 8cfm, 33% greater airflow within the hood and suit. The hood attaches to protective clothing with clips, for a secure fit. This system is designed for use in hazardous environments.
  • 320 degree unobstructed field of view
  • LiPo rechargeable battery lasts up to 12 hours
  • Ergonomically designed for comfort during long shifts and complex operations
  • One size fits all
  • Latex-free
The Sentinel XT® Clear PAPR System includes the S-3112 clear full hood, Sentinel XT® Non-Incendive Blower, 33" breathing tube, plug, LiPoMore Product Information