VWR® LED Digital Rotary Evaporators
Supplier: VWR International
76533-234 76533-258 76533-260 76533-262 76537-058
76533-234EA 4276.38 CAD
76533-234 76533-258 76533-260 76533-262 76537-058
VWR® LED Digital Rotary Evaporators
Evaporators Rotary Evaporators
This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional system in combination with a vacuum pump, vacuum controller and refrigerated circulator to provide excellent distillation solutions in many laboratory applications.
  • 5 L heating bath with a wide temperature range (ambient to 180 °C), independent temperature control allows it to be used separately. Water/oil heating mode can only be changed via a switch
  • Manual lift combined with auxiliary lift for precise
More Product Information