Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mills
Supplier: SPEX
8550-115 8550-230 8501-H 8504-H 8505-H 8506-H 8521-H
76533-058EA 37263.57 CAD
76533-058 76533-060 76533-154 76533-156 76533-158 76533-160 76533-162
Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mills
Mills
Grind your samples with minimal loss in a soundproof enclosure.
  • Store up to 10 different grinding programs
  • Robust IP 65 rated industrial interface
  • Accommodates various grinding dishes
  • Sound-insulated steel cabinet
  • Electronically controlled, fully reproducible grinding methods with Ethernet capabilities
The Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mill can crush and grind many types and sizes of samples with minimal sample loss and increased safety. It features an automated locking dish and guided dish positioning with an automaticMore Product Information