Cornerstone Lab Benches
Supplier: Treston
14-CS901-49 14-CS902-49 14-CS903-49 14-CS904-49 14-CS905-49 14-CS906-49 14-CS907-49 14-CS908-49 14-CS909-49 14-CS910-49 14-CS911-49 14-CS912-49 14-CS917-49 14-CS918-49 14-CS919-49 14-CS920-49 14-CS921-49 14-CS922-49 14-CS923-49 14-CS924-49 14-CS925-49 14-CS926-49 14-CS927-49 14-CS928-49 CSB3672-49 CSB3048-49 CSB3060-49 CSB3072-49 CSB3648-49 CSB3660-49 CSB2448-49 CSB2460-49 CSB2472-49
76532-894EA 3487.36 CAD
76532-894 76532-896 76532-898 76532-900 76532-902 76532-904 76532-906 76532-908 76532-910 76532-912 76532-914 76532-916 76532-918 76532-920 76532-922 76532-924 76532-926 76532-928 76532-930 76532-932 76532-934 76532-936 76532-938 76532-940 77575-708 77575-696 77575-698 77575-700 77575-704 77575-706 77575-688 77575-690 77575-692
Cornerstone Lab Benches
Furniture Tables and Workbenches
Treston® Cornerstone lab benches are manually height adjustable in 1" increments. All bench frames can be equipped with casters to create a mobile workstation solution.
  • Heavy duty steel structure supports loads up to 1500 lbs.
  • A cost-effective option
  • Wide range of accessories available to meet any need, including shelving, power, monitor arms, casters and more
  • QuickShip available
  • 10-year warranty
  • Paint color: Light neutral or snow white
  • Work surfaces: ³/₄" black phenolic or 1³/₁₆" black laminate (other options available upon request)
Treston® labMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable