Vikan® Split Bristle Angled Broom Head, Remco
Supplier: Remco Products
29162 29163 29164 29165 29166 29169
76528-722EA 57.89 CAD
76528-722 76528-724 76528-726 76528-728 76528-730 76528-732
Vikan® Split Bristle Angled Broom Head, Remco
Brooms
Split bristle angle head is designed to handle fine particles.
  • Soft/split bristles gently sweep away powders without leaving residues
  • Polypropylene block, polyester bristles
  • Euro (hygienic) thread type
Made of FDA compliant materials.