VHB™ 3M™ GPH-160GF Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-628PK 25.33 CAD
76524-628 76524-630 76524-632 76524-634 76524-636 76524-638 76524-640 76524-642 76524-644 76524-646 76524-648 76524-650 76524-652 76524-654 76524-656 76524-658 76524-660 76524-662 76524-664 76524-666 76524-668 76524-670 76524-672 76524-674 76524-676 76524-678 76524-680 76524-682 76524-684 76524-686 76524-688 76524-690 76524-692 76524-694 76524-696 76524-698 76524-700 76524-702 76524-704 76524-706 76524-708 76524-710 76524-712 76524-714 76524-716 76524-718 76524-720
VHB™ 3M™ GPH-160GF Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • Gray, 62 mil, 0.062" (1.6 mm) thick, acrylic adhesive with a conformable, acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives in appropriate applications
  • Maximum temperature: 450 °F (232 °C)
  • Adhesive strength: 19 lbs./inch
VHB™ tape GPH-160GF is double-sided acrylic foam tape with a red polyethylene film liner. TheMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable