VHB™ 3M™ 5952 Tapes, TapeCase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76524-018RL 15.1 CAD
76524-018 76524-020 76524-022 76524-024 76524-026 76524-028 76524-030 76524-032 76524-034 76524-036 76524-038 76524-040 76524-042 76524-044 76524-046 76524-048 76524-050 76524-052 76524-054 76524-056 76524-058 76524-060 76524-062 76524-064 76524-066 76524-068 76524-070 76524-072 76524-074 76524-076 76524-078 76524-080 76524-082 76524-084 76524-086 76524-088 76524-090 76524-092 76524-094 76524-096 76524-098 76524-100 76524-102 76524-104 76524-106 76524-108 76524-110 76524-112 76524-114 76524-116 76524-118 76524-120 76524-122 76524-124 76524-126 76524-128 76524-130 76524-132 76524-134 76524-136 76524-138 76524-140 76524-142 76524-144 76524-146 76524-148 76524-150 76524-152 76524-154 76524-156 76524-158 76524-160 76524-162 76524-164 76524-166 76524-168 76524-170 76524-172 76524-174 76524-176 76524-178 76524-180 76524-182 76524-184 76524-186 76524-188 76524-190 76524-192 76524-194 76524-196 76524-198 76524-200 76524-202 76524-204 76524-206 76524-208 76524-210 76524-212 76524-214 76524-216 76524-218 76524-220 76524-222 76524-224 76524-226 76524-228 76524-230 76524-232 76524-234 76524-236 76524-238 76524-240 76524-242 76524-244 76524-246 76524-248 76524-250 76524-252 76524-254 76524-256 76524-258 76524-260 76524-262 76524-264 76524-266 76524-268 76524-270 76524-272 76524-274 76524-276 76524-278 76524-280 76524-282 76524-284 76524-286 76524-288 76524-290 76524-292 76524-294 76524-296 76524-298 76524-300 76524-302 76524-304 76524-306 76524-308 76524-310 76524-312 76524-314 76524-316 76524-318 76524-320 76524-322 76524-324 76524-326 76524-328 76524-330 76524-332 76524-334 76524-336 76524-338 76524-340 76524-342 76524-344 76524-346 76524-348 76524-350 76524-352 76524-354 76524-356 76524-358 76524-360 76524-362 76524-364 76524-366 76524-368 76524-370 76524-372 76524-374 76524-376 76524-378 76524-380 76524-382 76524-384 76524-386 76524-388 76524-390 76524-392 76524-394 76524-396 76524-398 76524-400 76524-402 76524-404 76524-406 76524-408 76524-410 76524-412 76524-414 76524-416 76524-418 76524-420 76524-422 76524-424 76524-426 76524-428 76524-430 76524-432 76524-434 76524-436 76524-438 76524-440 76524-442 76524-444 76524-446 76524-448 76524-450 76524-452 76524-454 76524-456 76524-458 76524-460 76524-462 76524-464 76524-466 76524-468 76524-470 76524-472 76524-474 76524-476 76524-478 76524-480 76524-482 76524-484 76524-486 76524-488 76524-490 76524-492 76524-494 76524-496 76524-498 76524-500 76524-502 76524-504
VHB™ 3M™ 5952 Tapes, TapeCase
Tapes
VHB™ tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • Black, 45 mil thick, (1.1 mm), modified acrylic adhesive and very conformable acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welding, screws) or liquid adhesives
  • Maximum temperature: 300 °F (149 °C)
  • Adhesive strength: 22 lbs./inch
VHB™ tape 5952 is very conformable double-sided foam tape. This tape is designed for bonding with medium low surfaceMore Product Information