VHB™ 3M™ 4950 Tapes, Tapecase
Supplier: TAPECASE LTD. TE
8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11 8.88519E+11
76523-682RL 17.81 CAD
76523-682 76523-684 76523-686 76523-688 76523-690 76523-692 76523-694 76523-696 76523-698 76523-700 76523-702 76523-704 76523-706 76523-708 76523-710 76523-712 76523-714 76523-716 76523-718 76523-720 76523-722 76523-724 76523-726 76523-728 76523-730 76523-732 76523-734 76523-736 76523-738 76523-740 76523-742 76523-744 76523-746 76523-748 76523-750 76523-752 76523-754 76523-756 76523-758 76523-760 76523-762 76523-764 76523-766 76523-768 76523-770 76523-772 76523-774 76523-776 76523-778 76523-780 76523-782 76523-784 76523-786 76523-788 76523-790 76523-792 76523-794 76523-796 76523-798 76523-800 76523-802 76523-804 76523-806 76523-808 76523-810 76523-812 76523-814 76523-816 76523-818 76523-820 76523-822 76523-824 76523-826 76523-828 76523-830 76523-832 76523-834 76523-836 76523-838 76523-840 76523-842 76523-844 76523-846 76523-848 76523-850 76523-852 76523-854 76523-856 76523-858 76523-860 76523-862 76523-864 76523-866 76523-868 76523-870 76523-872 76523-874 76523-876 76523-878 76523-880 76523-882 76523-884 76523-886 76523-888 76523-890 76523-892 76523-894 76523-896 76523-898 76523-900 76523-902 76523-904 76523-906 76523-908 76523-910 76523-912 76523-914 76523-916 76523-918 76523-920 76523-922 76523-924 76523-926 76523-928 76523-930 76523-932 76523-934 76523-936 76523-938 76523-940 76523-942 76523-944 76523-946 76523-948 76523-950 76523-952 76523-954 76523-956 76523-958 76523-960 76523-962 76523-964 76523-966 76523-968 76523-970 76523-972 76523-974 76523-976 76523-978 76523-980 76523-982 76523-984 76523-986 76523-988 76523-990 76523-992 76523-994 76523-996 76523-998 76524-000 76524-002 76524-004 76524-006 76524-008 76524-010
VHB™ 3M™ 4950 Tapes, Tapecase
Tapes
VHB™ Tapes are fast and easy-to-use, with a permanent bonding method to provide high strength and long-term durability.
  • White, 1.1 mm (45 mil), general-purpose adhesive and firm acrylic foam core
  • Permanent bonding method provides high strength and long-term durability
  • Can replace mechanical fasteners (rivets, welds, screws) or liquid adhesives
  • Maximum temperature: 149 °C (300 °F)
  • Adhesive strength: 25 lbs./inch
VHB™ Tape 4950 replaces screws, rivets, and studs to permanently adhere one substrate to another while spreading the stress load across theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable