CyBio® Selma Semi-Automatic Pipetes 96/384
Supplier: Analytik Jena US
OL7001-26-211 OL7001-26-212 OL7001-26-213 OL7001-26-214 OL7001-26-216 OL7001-26-217
76521-978EA 32980.26 CAD
76521-978 76521-980 76521-982 76521-984 76521-986 76521-988
CyBio® Selma Semi-Automatic Pipetes 96/384
Automated Liquid Handling Systems
Precise and semi-automatic pipettor with 96 or 384 pipetting channels for easy and fast filling of 96- and 384-well microplates without the use of a computer. Applications like replication and reformatting, medium exchange as well as serial dilution can be executed reproducible and convenient.
  • Fast and precise processing of 96- and 384-well microplates without the need for programming
  • Compact footprint and fits on any lab bench as well as into most laminar flow hoods, Simple and intuitive operation via user guided methods
  • Easy and fast tip changing with
More Product Information