SP Bel-Art Reusable Silica Gel Desiccant Cartridges, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42049-0100
76521-464PK 134.28 CAD
76521-464
SP Bel-Art Reusable Silica Gel Desiccant Cartridges, Bel-Art Products, a part of SP
Desiccators Desiccants
Useful anywhere desiccation is needed, these cartridges contain inert, non-corrosive, and non-toxic silica gel.
  • Clear polypropylene dish allows instant visual inspection of silica gel, which changes from blue to pale pink as it absorbs moisture
  • Highly porous mesh membrane covers the dish providing excellent moisture absorption
  • Desiccant can be easily regenerated by heating cartridges in a 50 °C (122 °F) oven overnight, or at 100 °C (212 °F) for 6 hours
  • For more rapid drying, remove the membrane lid, transfer the spent gel crystals into a glass or porcelain
More Product Information