SHOWA CS721 Chemical Resistant Gloves
Supplier: Showa
CS721L-09 CS721M-08 CS721S-07 CS721XL-10 CS721XXL-11
76521-176BG 92.26 CAD
76521-176 76521-174 76521-172 76521-178 76521-180
SHOWA CS721 Chemical Resistant Gloves
Gloves
The SHOWA CS721 chemical-resistant gloves keep wearers safe from harmful substances, with an elongated forearm for extra protection.
  • Seamless knit, increased dexterity
  • Oil and chemical-resistant
  • Forearm protection
  • Anti-slip grip, robust durability
  • Double coated nitrile
  • Polyester liner, rough grip
A rough finish nitrile palm coating offers enhanced grip, while the powdered liner wicks away perspiration to prevent slipping and discomfort. The durable CS721 also keeps oil and dust out of the glove, ensuring optimum tactile precision.