Accessories for Corepoint™ Scientific Blood Bank Refrigerators and Plasma Freezer Units, Horizon Scientific
Supplier: Horizon Scientific
ANCHORKIT NSBB&PBALLASTKIT NSBB&PGLYCEROLKIT NSBB&PIOPQ
76520-784EA 217.38 CAD
76520-784 76520-786 76520-788 76520-794
Accessories for Corepoint™ Scientific Blood Bank Refrigerators and Plasma Freezer Units, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These additional accessories are designed for use with Corepoint™ Scientific blood bank refrigerators and plasma freezers.
  • Factory fitted options available are:
  • The anchor bracket kit is designed to add extra anchoring support
  • The upper solid ballast kit is an optional kit to replace the standard upper glycol bottle
  • The lower glycerol bottle kit is an optional kit to replace the standard lower solid ballast
The optional IQ/OQ/PQ add on for use with Corepoint™ Scientific blood bank refrigerators and plasma freezers should be ordered at the same time asMore Product Information