DuPont™ Tyvek® Autoclavable Bowl Cover, with Steam Indicator, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BWLT2A30-2PLY6 BGT2A0103E
76520-568CS 2398.44 CAD
76520-568 76918-244
DuPont™ Tyvek® Autoclavable Bowl Cover, with Steam Indicator, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • 6" 2-ply edges resist punctures
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator
  • Autoclavable at 121 to 125 °C
  • Hydrophobic
  • Lint-free
  • Manufactured and packaged in a controlled cleanroom environment
Stopper bowl covers provide a convenient,More Product Information