PIPETMAX® Software, Gilson
Supplier: GILSON, INC.
32000341 32000340 32000331 32000332 32000343
76518-592EA 6214.44 CAD
76518-592 76518-594 76518-596 76518-598 76518-600
PIPETMAX® Software, Gilson
Software Laboratory Software
TRILUTION® micro Software manages your automated PIPETMAX® workflows, allowing you to focus on more important tasks.
  • Easy to learn
  • Improved reproducibility and traceability for PCR and qPCR experiments
  • No more tedious normailization calculations
TRILUTION micro Software, the runtime control software for PIPETMAX, takes only minutes to learn, unlike other automated sample preparation solutions that require extensive training, or even dedicated technicians. From a single screen, TRILUTION micro lets you:
• Manage protocol managements, includingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable