Brady® SPC® Absorbents Oil-Only 14 gal. Drum Spill Kit, Brady
Supplier: Brady Worldwide
SKO-14
76518-282KT 271.31 CAD
76518-282
Brady® SPC® Absorbents Oil-Only 14 gal. Drum Spill Kit, Brady
Spill Control Materials
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructional guidelines so you're prepared when a spill occurs.
  • Absorption capacity 13 gal.
  • Oil Only spill control
White absorbents pick up petroleum-based fluids, paints and non-water-soluble chemicals while repelling water. Great for indoor and outdoor use. 14 gal. drum is a UN-approved, water- and chemical-resistant polyethylene container with lever-lock lid for small-medium response. Improve worker productivity and efficiency by placing spill kits around your facility close toMore Product Information