Brady® SPC® Absorbents ALLWIK Universal 14 gal. Drum Spill Kit, Brady
Supplier: Brady Worldwide
SKA-14
76518-278KT 264.49 CAD
76518-278
Brady® SPC® Absorbents ALLWIK Universal 14 gal. Drum Spill Kit, Brady
Spill Control Materials
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructional guidelines so you're prepared when a spill occurs.
  • Absorption capacity 12 gal.
  • Universal spill control
Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids. 14 gal. drum is a UN-approved, water- and chemical-resistant polyethylene container with lever-lock lid for small-medium response. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills. Spill kits areMore Product Information