VWR® Aloe-Coated Nitrile Gloves
Supplier: VWR International
76518-172 76518-174 76518-176 76518-178 76518-180
76518-172CS 458.17 CAD
76518-172 76518-174 76518-176 76518-178 76518-180
VWR® Aloe-Coated Nitrile Gloves
Gloves
The innovative aloe-coated nitrile glove is designed to combine the durability of nitrile with the soothing properties of aloe vera. The aloe coating provides a moisturizing and nourishing experience, keeping hands hydrated and preventing dryness and irritation.
  • Aloe-coated
  • 3.5 mil finger thickness
  • Powder-free
  • 9.5" length
  • Food contact compliant
  • High comfort and flexibility
  • Textured fingers
Perfect for exceptional grip and tactile sensitivity, ensuring optimal performance in various industries such as healthcare, laboratory, food handling, and more.