AMI Polymer Imaflow™ Platinum Cured Silicone Tubing, Foxx Life Sciences
Supplier: Foxx Life Sciences
FLO00300060025 FLO00300060050 FLO00300060100 FLO00300070025 FLO00300070050 FLO00300070100 FLO00300080025 FLO00300080050 FLO00300080100 FLO00320063025 FLO00320063050 FLO00320063100 FLO00400060025 FLO00400060050 FLO00400060100 FLO00400070025 FLO00400070050 FLO00400070100 FLO00400080025 FLO00400080050 FLO00400080100 FLO00400090025 FLO00400090050 FLO00400090100 FLO00400100025 FLO00400100050 FLO00400100100 FLO00480079025 FLO00480079050 FLO00480079100 FLO00480095025 FLO00480095050 FLO00480095100 FLO00500080025 FLO00500080050 FLO00500080100 FLO00500090025 FLO00500090050 FLO00500090100 FLO00500100025 FLO00500100050 FLO00500100100 FLO00500110025 FLO00500110050 FLO00500110100 FLO00600080025 FLO00600080050 FLO00600080100 FLO00600090025 FLO00600090050 FLO00600090100 FLO00600100025 FLO00600100050 FLO00600100100 FLO00600110025 FLO00600110050 FLO00600110100 FLO00600120025 FLO00600120050 FLO00600120100 FLO00630095025 FLO00630095050 FLO00630095100 FLO00630111025 FLO00630111050 FLO00630111100 FLO00630127025 FLO00630127050 FLO00630127100 FLO00790127025 FLO00790127050 FLO00790127100 FLO00800110025 FLO00800110050 FLO00800110100 FLO00800120025 FLO00800120050 FLO00800120100 FLO00800140025 FLO00800140050 FLO00800140100 FLO00800160025 FLO00800160050 FLO00800160100 FLO00900150025 FLO00900150050 FLO00900150100 FLO00900160025 FLO00900160050 FLO00900160100 FLO00950127025 FLO00950127050 FLO00950127100 FLO00950143025 FLO00950143050 FLO00950143100 FLO00950159025 FLO00950159050 FLO00950159100 FLO00950190025 FLO00950190050 FLO01000140025 FLO01000140050 FLO01000140100 FLO01000160025 FLO01000160050 FLO01000160100 FLO01000180025 FLO01000180050 FLO01200160025 FLO01200160050 FLO01200160100 FLO01200180025 FLO01200180050 FLO01200200025 FLO01200200050 FLO01270175025 FLO01270175050 FLO01270175100 FLO01270190025 FLO01270190050 FLO01270222050 FLO01270254050 FLO01500210050 FLO01590222050 FLO01590254050 FLO01600240050 FLO01900250050 FLO01900254050 FLO01900270050 FLO01900286050 FLO01900317050 FLO02500310050 FLO02540287050
76516-718PK 55.15 CAD
76516-718 76516-720 76516-722 76516-724 76516-726 76516-728 76516-730 76516-732 76516-734 76516-736 76516-738 76516-740 76516-742 76516-744 76516-746 76516-748 76516-750 76516-752 76516-754 76516-756 76516-758 76516-760 76516-762 76516-764 76516-766 76516-768 76516-770 76516-772 76516-774 76516-776 76516-778 76516-780 76516-782 76516-784 76516-786 76516-788 76516-790 76516-792 76516-794 76516-796 76516-798 76516-800 76516-802 76516-804 76516-806 76516-808 76516-810 76516-812 76516-814 76516-816 76516-818 76516-820 76516-822 76516-824 76516-826 76516-828 76516-830 76516-832 76516-834 76516-836 76516-838 76516-840 76516-842 76516-844 76516-846 76516-848 76516-850 76516-852 76516-854 76516-856 76516-858 76516-860 76516-862 76516-864 76516-866 76516-868 76516-870 76516-872 76516-874 76516-876 76516-878 76516-880 76516-882 76516-884 76516-886 76516-888 76516-890 76516-892 76516-894 76516-896 76516-898 76516-900 76516-902 76516-904 76516-906 76516-908 76516-910 76516-912 76516-914 76516-916 76516-918 76516-920 76516-922 76516-924 76516-926 76516-928 76516-930 76516-932 76516-934 76516-936 76516-938 76516-940 76516-942 76516-944 76516-946 76516-948 76516-950 76516-952 76516-954 76516-956 76516-958 76516-960 76516-962 76516-964 76516-966 76516-968 76516-970 76516-972 76516-974 76516-976 76516-978 76516-980 76516-982 76516-984
AMI Polymer Imaflow™ Platinum Cured Silicone Tubing, Foxx Life Sciences
Tubing Transfer Tubing
Imaflow™ is Platinum Grade Silicone Tubing designed for general purpose applications in within and outside of the Food and Pharmaceutical industries. Imaflow™ tubing is made from Pharmaceutical grade platinum cured silicone resin with a smooth bore finish compared to other peroxide grade tubing to reduce the risk of particle entrapment during fluid transfer.
  • Meets pharma grade requirement of ROW and Domestic markets
  • Greater Transparency and higher shelf life compare to other peroxide tubing
  • Free of heavy metals and Hazardous substances
  • Service Temperature
More Product Information