Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Supplier: Uniform Technology
CHPIN2-74WH-XS CHPIN2-74WH-S CHPIN2-74WH-M CHPIN2-74WH-L CHPIN2-74WH-XL CHPIN2-74WH2XL CHPIN2-74WH3XL
76515-930EA 26.7 CAD
76515-930 76515-932 76515-934 76515-936 76515-938 76515-940 76515-942
Reusable Altessa Grid Pull-Over Cleanroom Hoods with Face Mask, Uniform Technology
Workwear
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
  • Pull-over hood with built-in face mask
  • Vertical and horizontal snaps on back with elastic
Suitable for ISO Class 5 or higher cleanrooms.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Cleanroom
Risk area
Gender Unisex
Fabric 99% polyester/1% carbon