Test Tube Caps, Caplugs
Supplier: Caplugs
222-3812-02P 222-3812-02S 222-3817-02P 222-3817-02S 300-2908-020 300-2908-G20 300-2909-G20 300-2910-020 300-2911-020 300-2912-B20 300-2912-E20 300-2912-G20 300-2912-L20 300-2912-R20 300-2912-W20 300-2913-Y20 300-2916-020 300-2916-R20 300-2917-B2N 300-3812-02P 300-3812-02S 300-3817-02P 300-3817-02S 300-4708-020 300-4710-020 300-4712-020 300-4717-02N
76514-422CT 125.17 CAD
76514-422 76514-424 76514-426 76514-428 76514-430 76514-432 76514-434 76514-436 76514-438 76514-440 76514-442 76514-444 76514-446 76514-448 76514-450 76514-452 76514-454 76514-456 76514-458 76514-460 76514-462 76514-464 76514-466 76514-468 76514-470 76514-472 76514-474
Test Tube Caps, Caplugs
Closures Test/Sample Tube Closures
A variety of tube cap options to meet needs. Test Tube Caps are overfitting caps for use with culture tubes.
  • Two-position are only available in natural color
  • Standard test tube stopper caps are available in a wide variety of sizes and colors
  • Flat top stopper caps are designed to fit a variety of test tube sizes
Two-position caps are available non sterile or gamma irradiated for sterility.
Position one: Loose for aerobic culturing.
Position two: Tight for anaerobic culturing and leakproof sealing.
Order Now
SPECIFICATIONS
Environmentally Preferable