VWR® Standard Series Glass Door Laboratory Refrigerators
Supplier: VWR International
76514-260 76514-262 76514-264 76514-266 76514-268 76514-270 76514-272 76514-274 76514-276 76514-278
76514-260EA 8958.82 CAD
76514-260 76514-262 76514-264 76514-266 76514-268 76514-270 76514-272 76514-274 76514-276 76514-278
VWR® Standard Series Glass Door Laboratory Refrigerators
Refrigerators
These standard glass door laboratory refrigerators offer cold storage solutions at an economical pricepoint.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Adjustable shelving
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® standard series refrigerators utilize microprocessor temperature controllers, digital temperature displays, and temperature alarms. The are designed for multi-functional use in laboratory settings. All models also comeMore Product Information