VWR® Plus Laboratory Refrigerators, Stainless Steel
Supplier: VWR International
76514-202 76514-204 76514-206 76514-208 76514-210 76514-212
76514-202EA 8861.13 CAD
76514-202 76514-204 76514-206 76514-208 76514-210 76514-212
VWR® Plus Laboratory Refrigerators, Stainless Steel
Refrigerators
These VWR® Plus Series stainless steel refrigerators feature a multi-functional design for laboratory use.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Forced draft air circulation
  • Natural, hydrocarbon refrigerant
These VWR® refrigerators utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. All models are equipped with probe access ports, adjustable shelving,More Product Information