Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 4H1]

Supplier: Genscript
A01975-40
76509-976EA 243.16 CAD
76509-976
Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 4H1]
Antibodies
The product is specific for Bevacizumab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01976-40, Anti-Bevacizumab Antibody (6C3), mAb, Mouse.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Bevacizumab
Clonality: Monoclonal
Clone: 4H1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol Bevacizumab
Antigen name Bevacizumab
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 4H1
Reactivity
Host Mouse
Isotype IgG1
ELISA Yes
Immunogen Bevacizumab
Form Lyophilized
Purification Protein A affinity column
Concentration Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
Cross adsorption No
Storage buffer PBS, pH 7.4, containing 0.02% sodium azide
Storage temperature The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR