Humidity Test Chambers with Peltier Technology, KBF-S ECO, BINDER
Supplier: Binder
9020-0420 9020-0421 9020-0422 9020-0464
76509-742EA 26275.08 CAD
76509-742 76509-744 76509-746 76632-898
Humidity Test Chambers with Peltier Technology, KBF-S ECO, BINDER
Testing Chambers Humidity Chambers
The KBF-S ECO climatic chambers are very energy-efficient thanks to thermoelectric cooling technology with heat dissipation. The thermoelectric cooling technology means the KBF-S ECO series is also very quiet during operation.
  • Temperature range (max. 24 °C under ambient temperature): 0 to +70 °C
  • Humidity range: 10 to 80% RH, electrical: 120 V, 60 Hz, 1 phase
  • APT.line™ preheating chamber technology
  • Independent water supply via tank
  • LCD to display temperature and humidity along with additional information and alarms
  • Internal data logger, measured values can
More Product Information