Safety Waste Caps, SCAT
Supplier: SCAT AMERICAS, INC.
107034 107036 107037 107038 107050 107051 107052 107053 107054 107241 107242 107245 107246 107248 107256 107257 107258 107259 107913 107915 107917 107922 107924 107927 107928 107930 107935 107936 107942 107943 107945 107947 107949 107960 107962 107963 107965 107966 107967 107969 108023 108024 108025 108026 108030 108031 108046 108047 108055 108156 108200 108201 108205 108231 108407 307500 307912 307916 307918 307923 307925 307931 307944 307964 308401 308402 308403 308404 308921 308961 308964
76503-054EA 644.99 CAD
76503-054 76503-058 76503-060 76503-062 76503-080 76503-082 76503-084 76503-086 76503-088 76503-144 76503-146 76503-150 76503-152 76503-154 76503-160 76503-162 76503-164 76503-166 76503-316 76503-318 76503-320 76503-324 76503-326 76503-328 76503-330 76503-332 76503-336 76503-338 76503-346 76503-348 76503-350 76503-352 76503-354 76503-372 76503-374 76503-376 76503-378 76503-380 76503-382 76503-384 76503-420 76503-422 76503-424 76503-426 76503-432 76503-434 76503-452 76503-454 76503-462 76503-526 76503-552 76503-554 76503-556 76503-564 76503-580 76503-796 76503-808 76503-810 76503-812 76503-818 76503-820 76503-822 76503-824 76503-826 76503-834 76503-836 76503-838 76503-840 76503-842 76503-844 76503-846
Safety Waste Caps, SCAT
Closures Safety Caps and Safety Waste Caps
The SCAT Safety Waste Caps provide maximum safety for your HPLC liquid disposal. Safely dispose of your liquid laboratory waste without contaminating the ambient air by using SafetyWasteCaps with integrated exhaust air filters. These high-performance filters regulate the air pressure balance and prevent the escape of vapors due to the resulting overpressure in the disposal container. The variety of closures, connections and thread sizes allows optimal adaptation to your HPLC situation. Additional versions with grounding connections, level controls or safety funnels or offer every convenienceMore Product Information