SymLine® Tubes and Connectors, SCAT
Supplier: SCAT AMERICAS, INC.
106421 106426 106427 106430 106435 106436 106437 106438 106440 106449 106456 106457 106458 106471 106475 106476 106605 106608 106609 106611 107125
76502-884EA 264.11 CAD
76502-884 76502-888 76502-890 76502-892 76502-896 76502-898 76502-900 76502-902 76502-904 76502-908 76502-914 76502-916 76502-918 76502-922 76502-924 76502-926 76502-992 76502-994 76502-996 76502-998 76503-140
SymLine® Tubes and Connectors, SCAT
Fittings and Connectors
Place the collecting container/canister where needed; the pipe system can be exactly configured to suit specific needs.
  • Straight pipes are available in standard lengths of 300, 900 and 1200 mm
  • Angled pipes are available in the further lengths of 150 and 500 mm
  • A wide range of connectors, branches, and manifolds allow the pipe system to be connected precisely to the conditions in the laboratory
The SymLine® pipe system provides the connection to the central collection point.
Order Now
SPECIFICATIONS
Environmentally Preferable