Evergreen Compartmented Biopsy Cassette Tap with Cover, Caplugs Evergreen
Supplier: Caplugs
258-4264-W90 258-4279-W90
76502-842BG 287.7 CAD
76502-842 76502-844
Evergreen Compartmented Biopsy Cassette Tap with Cover, Caplugs Evergreen
Embedding Cassettes Biopsy Processing Cassettes
Caplugs Evergreen 2-, 4-, and 9-compartment cassettes with 0.6 mm holes help to customize biopsy processing with added efficiency and economy.
  • High density acetal construction
  • Acetal is resistant to histological solvents such as alcohol and xylene
  • Acetal will keep the specimen submerged
  • Lid and base preassembled
Modified ball-and-socket hinge design for ease of opening and closing and also for detachment.
Order Now
SPECIFICATIONS
Environmentally Preferable